Sodium acetate, anhydrous, ACS, 99.0% min
Sodium acetate, anhydrous, ACS, 99.0% min
Sodium acetate, anhydrous, ACS, 99.0% min
Thermo Scientific Chemicals

Sodium acetate, anhydrous, ACS, 99.0% min

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
製品番号(カタログ番号) 011554.A1
または、製品番号011554-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS127-09-3
IUPAC Namesodium acetate
Molecular FormulaC2H3NaO2
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=O
さらに表示
pHof a 5% solution 7.5-9.2 at 25?
Impurity contentCl 0.002%
Impurity contentFe 0.001%
Impurity contentPO₄ 0.001%
Impurity contentSO₄ 0.003%
さらに表示
Sodium acetate is usually used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography and it is also a plating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only