Potassium tetrafluoroborate, 99%
Potassium tetrafluoroborate, 99%
Potassium tetrafluoroborate, 99%
Thermo Scientific Chemicals

Potassium tetrafluoroborate, 99%

CAS: 14075-53-7 | BF4K | 125.90 g/mol
製品番号(カタログ番号) 011556.36
または、製品番号011556-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14075-53-7
IUPAC Namepotassium tetrafluoroboranuide
Molecular FormulaBF4K
InChI KeyAKEBROIVCDHVSD-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)F
さらに表示
Appearance (Color)White
FormCrystalline powder or chunks
Assay from Supplier's CofA≥98.5%
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as an additive in the alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only