Ammonium sulfate, ACS, 99.0% min
Ammonium sulfate, ACS, 99.0% min
Ammonium sulfate, ACS, 99.0% min
Thermo Scientific Chemicals

Ammonium sulfate, ACS, 99.0% min

In galvanizing steel, manufacturing of ammonia alum and sulfuric acid | CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
数量:
100 g
500 g
2 kg
10 kg
製品番号(カタログ番号) 011566.22
または、製品番号011566-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
さらに表示
Assay (unspecified)99.0% (NH₄)₂SO₄ min
Residue on ignition0.005%
Impurity contentCl 5ppm
Impurity contentPO₄ 5ppm
Heavy Metals(ICP-OES) 5ppm
さらに表示
In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. Ammonium sulfate is used in agriculture as fertilizer. It is a slow releasing nitrogenous fertilizer and a rich source of sulphur nutrient. It is employed as a dough conditioner in bread products. It finds use as raw materials for various chemical products such as dyes and pharmacetical products. It is a component of flame-proofing agents and fire extinguisher powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only