Thermo Scientific Chemicals

Ammonium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 011566.A9
10 kg, Each
Thermo Scientific Chemicals

Ammonium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 011566.A9
10 kg, Each
Quantity
Catalog number: 011566.A9
also known as 011566-A9
Price (USD)
Price: 256.00
Online price: 217.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7783-20-2
IUPAC Name
sulfuric acid diamine
Molecular Formula
H8N2O4S
InChI Key
BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES
N.N.OS(O)(=O)=O
Assay (unspecified)
99.0% (NH₄)₂SO₄ min
Residue on ignition
0.005%
Impurity content
Cl 5ppm
Impurity content
PO₄ 5ppm
Heavy Metals
(ICP-OES) 5ppm

Description

In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. Ammonium sulfate is used in agriculture as fertilizer. It is a slow releasing nitrogenous fertilizer and a rich source of sulphur nutrient. It is employed as a dough conditioner in bread products. It finds use as raw materials for various chemical products such as dyes and pharmacetical products. It is a component of flame-proofing agents and fire extinguisher powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text