Thermo Scientific Chemicals

Tin(IV) chloride, anhydrous, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 011571.57
4 x 500 g, Each
Thermo Scientific Chemicals

Tin(IV) chloride, anhydrous, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 011571.57
4 x 500 g, Each
Quantity
Catalog number: 011571.57
also known as 011571-57
Price (USD)
Price: 279.00
Online price: 237.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7646-78-8
IUPAC Name
tin(IV) chloride
Molecular Formula
Cl4H4Sn
InChI Key
HTGSJGUTBKFNJC-UHFFFAOYSA-N
SMILES
Cl.Cl.Cl.Cl.[Sn]
Total Metal Impurities
2% max.

Description

Tin(IV) chloride is a precursor to prepare organotin compounds such as tetralkyltin and dialkyldichlorotin(IV), which find applications as catalysts and polymer stabilizers. As a Lewis acid catalyst, it is used in Fridel-Crafts reactions for alkylation and cyclization. It is involved in the selective nitration of aromatic compounds in the presence of fuming nitric acid. Furthermore, it is used to prepare tin(IV) oxide coating by sol-gel process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text