Thermo Scientific Chemicals

Sodium hydrogen phosphate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011592.36
500 g, Each
Thermo Scientific Chemicals

Sodium hydrogen phosphate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011592.36
500 g, Each
Quantity
Catalog number: 011592.36
also known as 011592-36
Price (USD)
Price: 73.10
Online price: 62.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7782-85-6
IUPAC Name
disodium phosphoric acid heptahydrate
Molecular Formula
H17Na2O11P
InChI Key
PYLIXCKOHOHGKQ-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O
Assay (unspecified)
98.0-102.0% Na₂HPO₄.7H₂O min
Heavy Metals
(ICP-OES) 0.001%
Impurity content
Cl 0.001%
Impurity content
Fe 0.001%
Impurity content
SO₄ 0.005%

Description

Sodium hydrogen phosphate is widely used in detergents and cleaning agents. Combined with trisodium phosphate, it is employed in the food industry to adjust the pH and in water treatment to prevent calcium scale formation. It is utilized as a saline laxative to clean the bowel before a colonoscopy. It prevents the coagulation of condensed milk. It is also utilized as anti-caking additive in powdered products. It enhances the cook time and used as thickening agent in desserts and puddings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text