Potassium perchlorate, anhydrous, ACS, 99.0-100.5%
Potassium perchlorate, anhydrous, ACS, 99.0-100.5%
Potassium perchlorate, anhydrous, ACS, 99.0-100.5%
Thermo Scientific Chemicals

Potassium perchlorate, anhydrous, ACS, 99.0-100.5%

For Parr Bomb Work | CAS: 7778-74-7 | ClKO4 | 138.54 g/mol
製品番号(カタログ番号) 011630.22
または、製品番号011630-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7778-74-7
IUPAC Namepotassium perchlorate
Molecular FormulaClKO4
InChI KeyYLMGFJXSLBMXHK-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)(=O)=O
さらに表示
Assay (unspecified)99.0-100.5% KClO₄
Impurity contentFe 5ppm
Impurity contentCa 0.005%
Impurity contentCl 0.003%
Impurity contentNa 0.02%
さらに表示
For Parr Bomb WorkPotassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only