Lithium perchlorate, anhydrous, ACS, 95.0% min
Lithium perchlorate, anhydrous, ACS, 95.0% min
Lithium perchlorate, anhydrous, ACS, 95.0% min
Lithium perchlorate, anhydrous, ACS, 95.0% min
Lithium perchlorate, anhydrous, ACS, 95.0% min
Thermo Scientific Chemicals

Lithium perchlorate, anhydrous, ACS, 95.0% min

As an oxidizing agent | CAS: 7791-03-9 | ClH6LiO7 | 160.43 g/mol
数量:
10 g
100 g
500 g
製品番号(カタログ番号) 011639.22
または、製品番号011639-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7791-03-9
IUPAC Namelithium(1+) trihydrate perchlorate
Molecular FormulaClH6LiO7
InChI KeyJAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES[Li+].O.O.O.[O-][Cl](=O)(=O)=O
さらに表示
Impurity contentFe 5ppm
Impurity contentSO₄ 0.001%
pHof a 5% solution 6.0-7.5 at 25?
Heavy Metals(ICP-OES) 5ppm
Assay (unspecified)95.0% LiClO₄ min
さらに表示
Lithium perchlorate is used as an oxygen source for chemical oxygen generators, as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte, and act as chaotropic agent to denature proteins. It is used in the Baylis-Hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only