Thermo Scientific Chemicals
Ruthenium, 5% on alumina powder, Thermo Scientific Chemicals
Catalog number: 011749.14
25 g, Each
Thermo Scientific Chemicals
Ruthenium, 5% on alumina powder, Thermo Scientific Chemicals
Catalog number: 011749.14
25 g, Each
Quantity
Description
Hydrogenation of aliphatic carbonyls and aromatic rings, especially bulky molecules. Ruthenium on alumina catalyst plays an important role for copper-free sonogashira coupling reactions. It is likewise a useful catalyst for the racemization of secondary alcohols.