Ruthenium, 10% on activated carbon powder, reduced
Ruthenium, 10% on activated carbon powder, reduced
Ruthenium, 10% on activated carbon powder, reduced
Thermo Scientific Chemicals

Ruthenium, 10% on activated carbon powder, reduced

CAS: 7440-18-8 | Ru | 101.07 g/mol
製品番号(カタログ番号) 011757.14
または、製品番号011757-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
仕様
外見Black powder
CAS7440-18-8
ChEBICHEBI:30682
化学物質名または材質Ruthenium
Black
さらに表示

Ruthenium, 10% on activated carbon powder is used in hydrogenation of aromatic ring systems, particularly the hindered systems. It finds wide use for the reduction of aliphatic carbonyl compounds and carboxylic acids to alcohols, hydrogenation of anilines to cyclohexylamines, synthesis of ammonia, depolymerization of carbohydrates, Fischer-Tropsch synthesis, and hydrogenation of glucose to produce sorbitol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only