Sodium tetrachloropalladate(II) hydrate, 99.95% (metals basis), Pd 30%
Sodium tetrachloropalladate(II) hydrate, 99.95% (metals basis), Pd 30%
Sodium tetrachloropalladate(II) hydrate, 99.95% (metals basis), Pd 30%
Thermo Scientific Chemicals

Sodium tetrachloropalladate(II) hydrate, 99.95% (metals basis), Pd 30%

CAS: 13820-53-6 | Cl4Na2Pd | 294.20 g/mol
製品番号(カタログ番号) 011886.04
または、製品番号011886-04
価格(JPY)
-
数量:
2 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13820-53-6
IUPAC Namedisodium tetrachloropalladiumbis(ylium)
Molecular FormulaCl4Na2Pd
InChI KeyABKQFSYGIHQQLS-UHFFFAOYSA-J
SMILES[Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl
さらに表示
Total Metal Impurities0.05% max.
Assay (unspecified)Pd 30% min
Sodium tetrachloropalladate is used as a test salt for Pd allergy patch testing. It is used in chemical synthesis as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only