Potassium hexachloroiridate(III) hydrate, Ir 32.8% min
Thermo Scientific Chemicals

Potassium hexachloroiridate(III) hydrate, Ir 32.8% min

CAS: 35705-96-5
製品番号(カタログ番号) 011887.03
または、製品番号011887-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS35705-96-5
化学物質名または材質Potassium hexachloroiridate(III) hydrate
Concentration or Composition (by Analyte or Components)Ir 33.4% min.
EINECS番号237-854-0
式量522.22 (Anhydrous)
さらに表示
Potassium hexachloroiridate(III) hydrate is used to prepare other iridium compounds. It is used as a redox mediator to visualize human finger prints on a poly(vinylidene difluoride) membrane by scanning electrochemical microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only