Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Intermediate acidity. Suitable for chrome ore bearing samples up to 50% by weight. Often used in testing of glass. Samples must be fully oxidized
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Melting Point | 870°C |
Physical Form | Liquid |
Quantity | 1 kg |
Unit Size | Each |
Melting Point | 870°C |
Physical Form | Liquid |
Quantity | 5 x 1 kg |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
012087.A1 also known as 012087-A1 | Each | 1 kg | Price: 483.00 Online price: 410.65 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
012087.D9 also known as 012087-D9 | Each | 5 x 1 kg | Contact Us › | - | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|