Thermo Scientific Chemicals

Silver bromide, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 012110.06
5 g, Each
Thermo Scientific Chemicals

Silver bromide, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 012110.06
5 g, Each
Quantity
Catalog number: 012110.06
also known as 012110-06
Price (USD)
Price: 88.00
Online price: 74.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7785-23-1
IUPAC Name
silver(1+) bromide
Molecular Formula
AgBr
InChI Key
ADZWSOLPGZMUMY-UHFFFAOYSA-M
SMILES
[Br-].[Ag+]
Total Metal Impurities
0.002% max.

Description

Silver bromide is used in photographic film. Used in production of radiograph. It also used in infrared applications as windows and lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text