Thermo Scientific Chemicals

Ruthenium(III) nitrosylnitrate, Ru 31.3% min, Thermo Scientific Chemicals

Catalog number: 012175.06
5 g, Each
Thermo Scientific Chemicals

Ruthenium(III) nitrosylnitrate, Ru 31.3% min, Thermo Scientific Chemicals

Catalog number: 012175.06
5 g, Each
Quantity
Catalog number: 012175.06
also known as 012175-06
Price (USD)
Price: 360.00
Online price: 306.65
Your price:
Quantity
-

Chemical Identifiers

CAS
34513-98-9
IUPAC Name
λ²-ruthenium(2+) iminooxidanium trinitrate
Molecular Formula
N4O10Ru
InChI Key
WOSOOWIGVAKGOC-UHFFFAOYSA-N
SMILES
[Ru++].N#[O+].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Red/brown
Form
Crystalline
Assay (special)
Ruthenium, Ru: ≥31.3%

Description

Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text