Thermo Scientific Chemicals

Calcium zirconium oxide, 99.2% (metals basis), Thermo Scientific Chemicals

Catalog number: 012344.A1
1 kg, Each
Thermo Scientific Chemicals

Calcium zirconium oxide, 99.2% (metals basis), Thermo Scientific Chemicals

Catalog number: 012344.A1
1 kg, Each
Quantity
Catalog number: 012344.A1
also known as 012344-A1
Price (USD)

Chemical Identifiers

CAS
12013-47-7
IUPAC Name
calcium oxozirconiumbis(olate)
Molecular Formula
CaO3Zr
InChI Key
BJJVDFHQHSBAFC-UHFFFAOYSA-N
SMILES
[Ca++].[O-][Zr]([O-])=O
Total Metal Impurities
0.8% max.

Description

Calcium zirconium oxide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text