Barium titanium oxide, 99%
Barium titanium oxide, 99%
Barium titanium oxide, 99%
Thermo Scientific Chemicals

Barium titanium oxide, 99%

Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers | CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol
数量:
50 g
500 g
2 kg
製品番号(カタログ番号) 012348.18
または、製品番号012348-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12047-27-7
IUPAC Namebarium(2+) oxotitaniumbis(olate)
Molecular FormulaBaO3Ti
InChI KeyWNKMTAQXMLAYHX-UHFFFAOYSA-N
SMILES[Ba++].[O-][Ti]([O-])=O
さらに表示
Particle Size+325 Mesh Screen: ≤0.10%
Assay from Supplier's CofA(Ba+Sr)O: ≥65.0%
Composition(Ba+Sr)/Ti Ratio: 0.988-1.002
FormPowder
Assay from Supplier's CofATiO₂: ≥34.0%
Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only