Thermo Scientific Chemicals

Nickel ammonium sulfate hexahydrate, Thermo Scientific Chemicals

Catalog number: 012519.36
500 g, Each
Thermo Scientific Chemicals

Nickel ammonium sulfate hexahydrate, Thermo Scientific Chemicals

Catalog number: 012519.36
500 g, Each
Quantity
Catalog number: 012519.36
also known as 012519-36
Price (USD)
Price: 381.00
Online price: 329.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7785-20-8
Assay (unspecified)
98% min

Description

It is used in electroplating. Has limited use as dye mordants and is used in metal-finishing compositions. Nickel electrolyte for electroplating

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text