Thermo Scientific Chemicals

Potassium bromide, ACS, 99% min, Thermo Scientific Chemicals

Catalog number: 012612.30
250 g, Each
Thermo Scientific Chemicals

Potassium bromide, ACS, 99% min, Thermo Scientific Chemicals

Catalog number: 012612.30
250 g, Each
Quantity
Catalog number: 012612.30
also known as 012612-30
Price (USD)
Price: 55.10
Online price: 46.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7758-02-3
IUPAC Name
potassium bromide
Molecular Formula
BrK
InChI Key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES
[K+].[Br-]
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (unspecified)
99.0% KBr min
Heavy Metals
(ICP-OES) 5ppm
Impurity content
IO₃ 0.001%

Description

Used in manufacturing of photographic papers, engraving and lithography, in spectroscopy, and as an analytical lab reagent. Potassium bromide is applied as a veterinary drug, as an antiepileptic medication for dogs. It is utilized for the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for the efficient bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text