Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt)
Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt)
Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt)
Thermo Scientific Chemicals

Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt)

CAS: 12033-81-7 | H2N2O8PtS | 385.16 g/mol
数量:
∼10 g
∼0.5 g
∼2 g
製品番号(カタログ番号) 012627.NC
または、製品番号012627-NC
価格(JPY)
-
数量:
∼10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12033-81-7
IUPAC Namedihydrogen dinitroplatinumbis(ylium) sulfate
Molecular FormulaH2N2O8PtS
InChI KeySPPCQWREVYSZTJ-UHFFFAOYSA-N
SMILES[H+].[H+].[O-]S([O-])(=O)=O.[O-][N+](=O)[Pt++][N+]([O-])=O
さらに表示
Concentration (weight/weight)Pt 4-6%
Dihydrogen dinitrosulfatoplatinate(II) solution is used in the preparation of electrodeposition of platinum films and biocompatible platinum thin film electrodeposition on tin substrates, which acts as the interconnects material for implantable medical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only