Potassium hexabromoiridate(IV), Ir 25.4% min
Potassium hexabromoiridate(IV), Ir 25.4% min
Potassium hexabromoiridate(IV), Ir 25.4% min
Thermo Scientific Chemicals

Potassium hexabromoiridate(IV), Ir 25.4% min

CAS: 19121-78-9 | Br6IrK2 | 749.838 g/mol
製品番号(カタログ番号) 012651.03
または、製品番号012651-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS19121-78-9
IUPAC Namedipotassium hexabromoiridiumdiuide
Molecular FormulaBr6IrK2
InChI KeyRSDYABYRTHADQA-UHFFFAOYSA-H
SMILES[K+].[K+].Br[Ir--](Br)(Br)(Br)(Br)Br
さらに表示
Assay (unspecified)Ir 25.4% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only