Cesium nitrate, 99.8% (metals basis)
Cesium nitrate, 99.8% (metals basis)
Cesium nitrate, 99.8% (metals basis)
Thermo Scientific Chemicals

Cesium nitrate, 99.8% (metals basis)

Preparation of Cs compounds | CAS: 7789-18-6 | CsNO3 | 194.909 g/mol
製品番号(カタログ番号) 012884.30
または、製品番号012884-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7789-18-6
IUPAC Namecaesium(1+) nitrate
Molecular FormulaCsNO3
InChI KeyNLSCHDZTHVNDCP-UHFFFAOYSA-N
SMILES[Cs+].[O-][N+]([O-])=O
さらに表示
Total Metal Impurities0.2% max.
Preparation of cesium compoundsCesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only