Rubidium chloride, 99% (metals basis)
Rubidium chloride, 99% (metals basis)
Rubidium chloride, 99% (metals basis)
Thermo Scientific Chemicals

Rubidium chloride, 99% (metals basis)

Testing for perchloric acid, source of rubidium, catalyst | CAS: 7791-11-9 | ClRb | 120.92 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 012892.06
または、製品番号012892-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7791-11-9
IUPAC Namerubidium(1+) chloride
Molecular FormulaClRb
InChI KeyFGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES[Cl-].[Rb+]
さらに表示
Appearance (Color)White
FormPowder or crystals or crystalline powder
Total Metal Impurities≤1.0%
Appearance (Color)White
FormPowder or crystals or crystalline powder
さらに表示
It is used in testing for perchloric acid in laboratories, and is a source of rubidium. Rubidium chloride is used as a catalyst and additive in petrol. It has been employed to prepare molecular nanowires for nanoscale devices. Thin layer of rubidium chloride enhance the attributes of organic light-emitting diodes. Rubidium chloride also plays an essential role for enhancement in the memory storage property. It is an excellent non-invasive biomarker, and used in biomedical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only