Thermo Scientific Chemicals

Molybdenum(VI) oxide, Puratronic™, 99.9995% (metals basis excluding W), Thermo Scientific Chemicals

Catalog number: 012930.14
25 g, Each
Thermo Scientific Chemicals

Molybdenum(VI) oxide, Puratronic™, 99.9995% (metals basis excluding W), Thermo Scientific Chemicals

Catalog number: 012930.14
25 g, Each
Quantity
Catalog number: 012930.14
Price (USD)
Price: 122.00
Online price: 103.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1313-27-5
IUPAC Name
trioxomolybdenum
Molecular Formula
MoO3
InChI Key
JKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILES
O=[Mo](=O)=O
Total Metal Impurities excluding W
0.0005% max.

Description

It is used as a selective oxidation catalyst, and as a molybdenum oxidase enzyme. It is a source of molybdenum compounds and molybdenum metal which serves as an additive to steel and corrosion-resistant alloys. It is used for lubricants and lubricant additives. It has applications in process regulators, pigments, corrosion inhibitors, electrochemical devices, displays, and smoke suppressants. It finds use in analytical chemistry, ceramic glazes, enamels, energy storage, and in the manufacture of acrylonitrile through catalytic oxidation of ammonia and propene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text