Selenium(IV) chloride, 99.5% (metals basis)
Selenium(IV) chloride, 99.5% (metals basis)
Selenium(IV) chloride, 99.5% (metals basis)
Thermo Scientific Chemicals

Selenium(IV) chloride, 99.5% (metals basis)

CAS: 10026-03-6 | Cl4Se | 220.771 g/mol
製品番号(カタログ番号) 013096.14
または、製品番号013096-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10026-03-6
IUPAC Nametetrachloro-λ⁴-selane
Molecular FormulaCl4Se
InChI KeyLNBXMNQCXXEHFT-UHFFFAOYSA-N
SMILESCl[Se](Cl)(Cl)Cl
さらに表示
Total Metal Impurities0.5% max.
It is used in the synthesis of other selenium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only