Yttrium(III) carbonate hydrate, REacton™, 99.99% (REO)
Yttrium(III) carbonate hydrate, REacton™, 99.99% (REO)
Yttrium(III) carbonate hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Yttrium(III) carbonate hydrate, REacton™, 99.99% (REO)

CAS: 38245-39-5 | C3O9Y2 | 357.84 g/mol
製品番号(カタログ番号) 013157.14
または、製品番号013157-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS38245-39-5
IUPAC Namediyttrium(3+) tricarbonate
Molecular FormulaC3O9Y2
InChI KeyQVOIJBIQBYRBCF-UHFFFAOYSA-H
SMILES[Y+3].[Y+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
さらに表示
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only