Rubidium sulfate, 99% (metals basis)
Rubidium sulfate, 99% (metals basis)
Rubidium sulfate, 99% (metals basis)
Thermo Scientific Chemicals

Rubidium sulfate, 99% (metals basis)

CAS: 7488-54-2 | H2O4RbS | 183.54 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 013493.22
または、製品番号013493-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7488-54-2
IUPAC Namesulfuric acid rubidium
Molecular FormulaH2O4RbS
InChI KeyXNNUAZGCOYTNQE-UHFFFAOYSA-N
SMILES[Rb].OS(O)(=O)=O
さらに表示
Total Metal Impurities1% max.
Rubidium sulfate is used in hematology stain. It is also used as the raw materials for making kinds of Rubidium salts. It is fused on a ceramic bead and used as thermoionic detector. It is also used as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only