Copper single crystal disc, 10mm (0.39in) dia, 1-3mm (0.04-0.1in) thick, (111) orientation
Copper single crystal disc, 10mm (0.39in) dia, 1-3mm (0.04-0.1in) thick, (111) orientation
Copper single crystal disc, 10mm (0.39in) dia, 1-3mm (0.04-0.1in) thick, (111) orientation
Thermo Scientific Chemicals

Copper single crystal disc, 10mm (0.39in) dia, 1-3mm (0.04-0.1in) thick, (111) orientation

CAS: 7440-50-8 | Cu | 63.55 g/mol
製品番号(カタログ番号)数量
013573.KG
または、製品番号013573-KG
1 Ea.
製品番号(カタログ番号) 013573.KG
または、製品番号013573-KG
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
仕様
CAS7440-50-8
IUPAC名copper
InChIキーRYGMFSIKBFXOCR-UHFFFAOYSA-N
MDL番号MFCD00010965
分子式Cu
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.