Iron(II) iodide, ultra dry, 99.99% (metals basis)
Iron(II) iodide, ultra dry, 99.99% (metals basis)
Iron(II) iodide, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Iron(II) iodide, ultra dry, 99.99% (metals basis)

Light and Air Sensitive and Hygroscopic | CAS: 7783-86-0 | FeI2
製品番号(カタログ番号) 013619.03
または、製品番号013619-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7783-86-0
IUPAC Nameλ²-iron(2+) diiodide
Molecular FormulaFeI2
InChI KeyBQZGVMWPHXIKEQ-UHFFFAOYSA-L
SMILES[Fe++].[I-].[I-]
さらに表示
Residual water< 100 ppm
Total Metal Impurities0.01% max.
Particle Size-10 Mesh
Appearance (Color)Red-brown
FormBeads
Iron(II) iodide is used as a catalyst in organic reactions. It is also used as a compound in semiconductors and other high purity applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only