Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

CAS: 13708-63-9 | F3Tb | 215.92 g/mol
製品番号(カタログ番号) 013657.14
または、製品番号013657-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13708-63-9
IUPAC Nameterbium(3+) trifluoride
Molecular FormulaF3Tb
InChI KeyLKNRQYTYDPPUOX-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Tb+3]
さらに表示
Total Rare Earth Oxide Impurities0.1% max.
Terbium(III) fluoride is used as a source of terbium. It is also useful in the preparation of fluoride glasses. Further, it is used in the preparation of electroluminescent thin films and luminescent zinc sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only