Neodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Neodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Neodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

CAS: 13709-42-7 | F3Nd | 201.24 g/mol
製品番号(カタログ番号) 013661.14
または、製品番号013661-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13709-42-7
IUPAC Nameneodymium(3+) trifluoride
Molecular FormulaF3Nd
InChI KeyXRADHEAKQRNYQQ-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Nd+3]
さらに表示
Total Rare Earth Oxide Impurities0.1% max.
Neodymium(III) fluoride, anhydrous is used extensively in the preparation and study of fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only