Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL
Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL
Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL
Thermo Scientific Chemicals
Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL
CAS: 7440-02-0 | Ni | 58.69 g/mol
数量:
50 mL
100 mL
500 mL
製品番号(カタログ番号) 013839.AP
または、製品番号013839-AP
価格(JPY)
-
数量:
500 mL
一括またはカスタム形式をリクエストする
仕様
CAS7440-02-0
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:28112
化学物質名または材質Nickel
さらに表示
Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.