Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)

CAS: 7440-00-8 | Nd | 144.24 g/mol
数量:
25 x 25 mm
50 x 50 mm
製品番号(カタログ番号) 013964.FI
または、製品番号013964-FI
価格(JPY)
-
見積もりを依頼する
数量:
50 x 50 mm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-00-8
IUPAC Nameneodymium
Molecular FormulaNd
InChI KeyQEFYFXOXNSNQGX-UHFFFAOYSA-N
SMILES[Nd]
さらに表示
Total Rare Earth Oxide Impurities0.1% max.
Neodymium applications include lasers, glass coloring and tinting, dielectrics and, most importantly, as the fundamental basis for neodymium-iron-boron (Nd2Fe14B) permanent magnets. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only