Platinum(IV) sulfide, Pt 74.8% min
Platinum(IV) sulfide, Pt 74.8% min
Platinum(IV) sulfide, Pt 74.8% min
Thermo Scientific Chemicals

Platinum(IV) sulfide, Pt 74.8% min

CAS: 12038-21-0 | PtS2 | 259.20 g/mol
製品番号(カタログ番号) 013992.03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12038-21-0
IUPAC Namedisulfanylideneplatinum
Molecular FormulaPtS2
InChI KeyRKNFJIIYAUSTJA-UHFFFAOYSA-N
SMILESS=[Pt]=S
さらに表示
Assay (unspecified)Pt 74.8% min
Ultra high purity and high purity compositions of platinum sulfide improve both optical quality and usefulness as scientific standards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only