Ruthenium powder, -325 mesh, 99.9% (metals basis)
Ruthenium powder, -325 mesh, 99.9% (metals basis)
Ruthenium powder, -325 mesh, 99.9% (metals basis)
Thermo Scientific Chemicals

Ruthenium powder, -325 mesh, 99.9% (metals basis)

CAS: 7440-18-8 | Ru | 101.07 g/mol
製品番号(カタログ番号) 013994.04
または、製品番号013994-04
価格(JPY)
-
数量:
2 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-18-8
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
さらに表示
FormPowder, -325 mesh
Total Metal Impurities≤0.1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only