Titanium rod, 3.175mm (0.125in) dia, 99.99% (metals basis)
Titanium rod, 3.175mm (0.125in) dia, 99.99% (metals basis)
Titanium rod, 3.175mm (0.125in) dia, 99.99% (metals basis)
Thermo Scientific Chemicals

Titanium rod, 3.175mm (0.125in) dia, 99.99% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol
数量:
5 cm
25 cm
4 x 25 cm
製品番号(カタログ番号) 014000.BQ
または、製品番号014000-BQ
価格(JPY)
-
数量:
5 cm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
さらに表示
FormRod, 3.175mm (0.125in) diameter.
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.99% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only