Ammonium bromide, ACS, 99.0% min
Ammonium bromide, ACS, 99.0% min
Ammonium bromide, ACS, 99.0% min
Ammonium bromide, ACS, 99.0% min
Thermo Scientific Chemicals

Ammonium bromide, ACS, 99.0% min

CAS: 12124-97-9 | BrH4N | 97.94 g/mol
数量:
25 g
100 g
500 g
2 kg
製品番号(カタログ番号) 014257.14
または、製品番号014257-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12124-97-9
IUPAC Nameammonium bromide
Molecular FormulaBrH4N
InChI KeySWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES[NH4+].[Br-]
さらに表示
Appearance (Color)White
FormSolid
Assay (unspecified)≤99.0% NH4Br
Barium content≤0.002%
Chloride content≤0.2%
さらに表示
Corrosion inhibitors, manufacturing of photographic films, plates, paper, lithography, fireproofing of woodsAmmonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only