Thermo Scientific Chemicals

Sodium azide, 99%, Thermo Scientific Chemicals

Catalog number: 014314.22
100 g, Each
Thermo Scientific Chemicals

Sodium azide, 99%, Thermo Scientific Chemicals

Catalog number: 014314.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 014314.22
also known as 014314-22
Price (USD)
Price: 62.50
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
26628-22-8
IUPAC Name
sodium azide
Molecular Formula
N3Na
InChI Key
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES
[Na+].[N-]=[N+]=[N-]
Loss on Drying
≤0.1%
Form
Powder
Comment
Alkalinity (as NaOH) 0.5% max
Assay from Supplier's CofA
≥98.5%
Insoluble material
≤500ppm

Description

In airbag inflation, as a preservative in diagnostic medicinals.Sodium azide is used in organic synthesis for the introduction of an azide group to prepare a desired compound. It acts as a precursor to prepare inorganic azides like lead azide and silver azide. In biochemistry, it is useful as a probe reagent and a preservative. It also acts as a nitridizing agent and cytochrome oxidase inhibitor. Further, it is used as a propellant in airbags.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text