Thermo Scientific Chemicals

Thulium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014582.09
10 g, Each
Thermo Scientific Chemicals

Thulium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 014582.09
10 g, Each
Quantity
Catalog number: 014582.09
also known as 014582-09
Price (USD)
Price: 292.00
Online price: 248.65
Your price:
Quantity
-

Chemical Identifiers

CAS
314041-04-8
IUPAC Name
thulium(3+) triacetate
Molecular Formula
C6H9O6Tm
InChI Key
SNMVVAHJCCXTQR-UHFFFAOYSA-K
SMILES
[Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Appearance (Color)
White to pale yellow or pale green
Assay from Suppliers CofA
Total Rare Earth Oxide Impurities (REO): ≤0.1%
Form
Crystals or powder or crystalline powder or crystalline aggregates

Description

Thulium Acetate is the important dopant for silica-based fiber amplifiers. It can also be used in portable X-ray devices that has been bombarded in a nuclear reactor as a radiation source. Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. The other main application is used for optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text