Matrix oil, 75 centistoke hydrocarbon oil
Thermo Scientific Chemicals

Matrix oil, 75 centistoke hydrocarbon oil

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
製品番号(カタログ番号) 014621.LV
または、製品番号014621-LV
価格(JPY)
-
数量:
1 gal.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS8042-47-5
CommentCOA includes trace metal concentrations.
CommentBlank
Viscosity75 centistoke
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only