Manganese(II) oxalate dihydrate, Mn 30% min
Manganese(II) oxalate dihydrate, Mn 30% min
Manganese(II) oxalate dihydrate, Mn 30% min
Thermo Scientific Chemicals

Manganese(II) oxalate dihydrate, Mn 30% min

As a paint and varnish drier | CAS: 6556-16-7 | C2H4MnO6 | 178.986 g/mol
数量:
5 g
50 g
250 g
1 kg
製品番号(カタログ番号) 014685.06
または、製品番号014685-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6556-16-7
IUPAC Namemanganese(2+) dihydrate oxalate
Molecular FormulaC2H4MnO6
InChI KeyHDJUVFZHZGPHCQ-UHFFFAOYSA-L
SMILESO.O.[Mn++].[O-]C(=O)C([O-])=O
さらに表示
FormCrystals or powder or crystalline powder
Assay from Suppliers CofAManganese, Mn: ≥30%
Appearance (Color)White to pale pink
Manganese(II) oxalate dihydrate is used as a paint and varnish drier. It is also used as a chemical reagent as well as to prepare other manganese compounds. In addition, it is used as a semiconductor photosensitive material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only