Styrene, 99.5% stab. with 4-tert-butylcatechol
Styrene, 99.5% stab. with 4-tert-butylcatechol
Styrene, 99.5% stab. with 4-tert-butylcatechol
Styrene, 99.5% stab. with 4-tert-butylcatechol
Thermo Scientific Chemicals

Styrene, 99.5% stab. with 4-tert-butylcatechol

CAS: 100-42-5 | C8H8 | 104.15 g/mol
数量:
50 mL
250 mL
1 L
製品番号(カタログ番号) 016199.AK
または、製品番号016199-AK
価格(JPY)
-
見積もりを依頼する
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS100-42-5
IUPAC Nameethenylbenzene
Molecular FormulaC8H8
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1
さらに表示
Assay from Supplier's CofA≥99.45% (GC)
CommentAldehydes as benzaldehyde: ≤100 ppm
Impurity contentEthylbenzene: ≤500 ppm
Appearance (Color)Colorless
CommentPolymer: ≤10 ppm
さらに表示
Styrene is used in the preparation of polystyrene, which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include in refrigerator liners, in medical devices and in small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN) which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only