Tetraethylammonium hydroxide, 25% w/w in methanol
Tetraethylammonium hydroxide, 25% w/w in methanol
Tetraethylammonium hydroxide, 25% w/w in methanol
Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 25% w/w in methanol

CAS: 77-98-5 | C8H21NO | 147.26 g/mol
製品番号(カタログ番号) 016328.30
または、製品番号016328-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS77-98-5
IUPAC Nametetraethylazanium hydroxide
Molecular FormulaC8H21NO
InChI KeyLRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES[OH-].CC[N+](CC)(CC)CC
さらに表示
Assay from Supplier's CofA≥24.0 to ≤26.0% (titration)
Proton NMRConforms
Appearance (Color)Clear colorless to pale yellow
FormLiquid
It is used as a phase transfer catalyst, the template agent synthesized by molecular sieve and cleaning agents, as a surfactant, used to anisotropically etch silicon, in thermochemolysis, as a basic solvent in the development of acidic photo resist in the photolithography process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only