Iron(II) sulfide, 99% (metals basis)
Iron(II) sulfide, 99% (metals basis)
Iron(II) sulfide, 99% (metals basis)
Iron(II) sulfide, 99% (metals basis)
Thermo Scientific Chemicals

Iron(II) sulfide, 99% (metals basis)

CAS: 1317-37-9 | FeS | 87.91 g/mol
製品番号(カタログ番号) 017422.18
または、製品番号017422-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1317-37-9
IUPAC Nameλ²-iron(2+) sulfanediide
Molecular FormulaFeS
InChI KeyGNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES[S--].[Fe++]
さらに表示
Total Metal Impurities1% max.
Iron(II) sulfide is used to generate hydrogen sulfide instantaneously by reacting with hydrochloric acid. In laboratory, Kipp’s apparatus is utilized for this purpose. It is used as a re-sulphurizing and alloying agent, as a reducing agent to remove heavy metal impurities from phosphoric acid. It is used to control hydrogen embrittlement in alloy and stainless steel industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only