Tetrafluoroboric acid-diethyl ether complex, 50-55% w/w HBF4
Tetrafluoroboric acid-diethyl ether complex, 50-55% w/w HBF4
Tetrafluoroboric acid-diethyl ether complex, 50-55% w/w HBF4
Thermo Scientific Chemicals

Tetrafluoroboric acid-diethyl ether complex, 50-55% w/w HBF4

CAS: 67969-82-8 | C4H11BF4O | 161.935 g/mol
製品番号(カタログ番号) 018208.AK
または、製品番号018208-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67969-82-8
IUPAC Nameboron(3+) hydrogen ethoxyethane tetrafluoride
Molecular FormulaC4H11BF4O
InChI KeyGYFCYMJGLCBLMX-UHFFFAOYSA-K
SMILES[H+].[B+3].[F-].[F-].[F-].[F-].CCOCC
さらに表示
Assay (unspecified)50 to 57 wt% HBF4
Appearance (Color)Colorless to brown
FormLiquid
Tetrafluoroboric acid-diethyl ether acts as an effective catalyst used for various carbohydrate protection reactions. It is also used as a catalyst in the preparation of 4-fluoropiperidines by reaction between aldehydes and N-tosyl homoallylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only