Copper(II) methoxide, typically 98% (metals basis)
Thermo Scientific Chemicals

Copper(II) methoxide, typically 98% (metals basis)

CAS: 1184-54-9
製品番号(カタログ番号) 018861.14
または、製品番号018861-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1184-54-9
CAS1184-54-9
Total Metal Impurities≤2.0%
Appearance (Color)Blue to green
FormPowder or crystals or crystalline powder
Appearance (Color)Blue to green
FormPowder or crystals or crystalline powder
さらに表示
Copper(II) methoxide is used as primary and secondary intermediates. Treatment of these methoxide derivatives with excess water results in the clean formation of mononuclear copper(II) hydroxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only