Hexanes, mixed isomers, (60+% n-hexane)
Hexanes, mixed isomers, (60+% n-hexane)
Hexanes, mixed isomers, (60+% n-hexane)
Thermo Scientific Chemicals

Hexanes, mixed isomers, (60+% n-hexane)

CAS: 92112-69-1 | C6H14 | 86.18 g/mol
数量:
500 mL
1 L
4 L
4 x 1 L
製品番号(カタログ番号) 019353.M1
または、製品番号019353-M1
価格(JPY)
-
数量:
4 x 1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS92112-69-1
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
さらに表示
FormLiquid
Assay (GC)≥99.5% as C6 Hydrocarbons [US]
Optical Absorbance at195 nm ≤1.000 [US]
Optical Absorbance at250 nm ≤0.020 [US]
Residue on Evaporation≤3 ppm
さらに表示
Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only