Lithium tetrachlorocuprate, 0.1M in THF
Lithium tetrachlorocuprate, 0.1M in THF
Lithium tetrachlorocuprate, 0.1M in THF
Lithium tetrachlorocuprate, 0.1M in THF
Thermo Scientific Chemicals

Lithium tetrachlorocuprate, 0.1M in THF

CAS: 15489-27-7 | Cl4CuLi2 | 219.226 g/mol
数量:
100 mL
500 mL
製品番号(カタログ番号) 020620.AP
または、製品番号020620-AP
価格(JPY)
-
見積もりを依頼する
数量:
500 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS15489-27-7
IUPAC Namecopper(2+) dilithium(1+) tetrachloride
Molecular FormulaCl4CuLi2
InChI KeyHCJWWBBBSCXJMS-UHFFFAOYSA-J
SMILES[Li+].[Li+].[Cl-].[Cl-].[Cl-].[Cl-].[Cu++]
さらに表示
FormLiquid
Appearance (Color)Orange to dark orange
Assay from Supplier's CofA0.09-0.12M (titration by AgNO₃)
Assay from Supplier's CofA0.09-0.12M (titration by Na₂S₂O₃)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only