Trichloroacetic acid, ACS, 99% min
Trichloroacetic acid, ACS, 99% min
Trichloroacetic acid, ACS, 99% min
Trichloroacetic acid, ACS, 99% min
Thermo Scientific Chemicals

Trichloroacetic acid, ACS, 99% min

Protein precipitation reagent | CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol
数量:
50 g
250 g
1 kg
製品番号(カタログ番号) 022156.18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS76-03-9
IUPAC Nametrichloroacetic acid
Molecular FormulaC2HCl3O2
InChI KeyYNJBWRMUSHSURL-UHFFFAOYSA-N
SMILESOC(=O)C(Cl)(Cl)Cl
さらに表示
Impurity contentSulfate (SO₄): ≤0.02%
Insoluble material≤0.01%
Heavy Metals (as Pb)≤0.002%
Appearance (Color)Colorless to white
FormSolid
さらに表示
Protein precipitation reagentTrichloroacetic acid is used as a precipitating agent in biochemistry for precipitation of proteins, DNA and RNA. It is an active ingredient used in cosmetic treatments like chemical peels, tattoo removal and the treatment of warts including genital warts. It is also used to determine protein concentration and as a decalcifier and fixative in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only