Calcium iodide, anhydrous, 99.5% (metals basis)
Calcium iodide, anhydrous, 99.5% (metals basis)
Calcium iodide, anhydrous, 99.5% (metals basis)
Calcium iodide, anhydrous, 99.5% (metals basis)
Thermo Scientific Chemicals

Calcium iodide, anhydrous, 99.5% (metals basis)

Light Sensitive and Hygroscopic | CAS: 10102-68-8 | CaI2 | 293.887 g/mol
製品番号(カタログ番号) 022395.14
または、製品番号022395-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10102-68-8
IUPAC Namecalcium diiodide
Molecular FormulaCaI2
InChI KeyUNMYWSMUMWPJLR-UHFFFAOYSA-L
SMILES[Ca++].[I-].[I-]
さらに表示
Total Metal Impurities0.5% max.
Calcium iodide, anhydrous is used as a drug in upper respiratory combinations, expectorants. And it is also used as Laboratory chemicals, Manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only